Learn More
Apexbio Technology LLC LY2606368 1234015-52-1 25mg
SDP
Supplier:  Apexbio Technology LLC B108825
Encompass_Preferred
LY2606368 (CAS 1234015-52-1) is a selective ATP-competitive inhibitor of checkpoint kinase 1 (CHK1) a serine/threonine kinase integral to the DNA damage response pathway In SW1990 pancreatic cancer cells LY2606368 exhibits potent inhibition of cell proliferation with an IC50 value of 1 5 nM The compound suppresses CHK1 autophosphorylation and induces phosphorylation of H2AX reflective of DNA damage signaling In vivo studies show that LY2606368 reduces tumor growth in SW1990 xenograft and SKOV3 orthotopic models and lowers tumor metastasis rates Due to low oral bioavailability administration is intravenous LY2606368 is under clinical investigation as an anticancer agent
Catalog No. 50-000-89601
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.