Learn More
Selleck Chemical LLC Sotagliflozin
SDP
Supplier:  Selleck Chemical LLC S81031G
Encompass_Preferred
Sotagliflozin is an oral dual SGLT1/SGLT2 inhibitor with IC50 of 36 nM and 1 8 nM respectively Phase 3
Catalog No. 50-000-00342
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.