Learn More
Agilent Technologies AdvanceBio Sialidase S, His-tagged
Supplier:  Agilent Technologies GK80021
Encompass
Enzyme releases alpha(2-3)-linked sialic acid from oligosaccharides, glycoproteins, complex carbohydrates. Includes 5x reaction buffer B (250 mM sodium phosphate pH 6.0).
Catalog No. NC1154755
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.