Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyoxal-bis(2-hydroxyanil), ≥97.0% (from N), Solstice

Catalog No. 60046229 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
6 x 25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-229 25 g Glass Bottle
60-046-230 6 x 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-229 Supplier Solstice Supplier No. 3325525G
Add to Cart
Add to Cart

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1149-16-2
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00002183
InChI Key OMJNMRJMNUREOJ-KOBPNRPCSA-N
PubChem CID 5356911
SMILES OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O

Specifications

Melting Point 203°C to 211°C
UN Number NONH for all modes of transport
Beilstein 2808815
IUPAC Name 6-[(E)-2-(2-hydroxyanilino)ethenyl]iminocyclohexa-2,4-dien-1-one
Formula Weight 240.26g/mol
Percent Purity ≥97% (from N)
Odor Odourless
Packaging Glass Bottle
Suitability Passes test for suitable for metal definition
Color Yellow
Quantity 25 g
Chemical Name or Material Glyoxal-bis(2-hydroxyanil)
Show More Show Less
RTECSNumber 214-560-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.