Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Glyoxal, pure, 40 wt.% solution in water

Catalog No. AC156220025
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156220025 2.5 L Glass bottle
AC156225000 500 mL Glass bottle
AC156220010 1 L Glass bottle
AC156220250 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC156220025 Supplier Thermo Scientific Chemicals Supplier No. 156220025
Add to Cart

CAS: 107-22-2 | C2H2O2 | 58.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glyoxal
Name Note 40 wt.% Solution in Water
CAS 107-22-2, 7732-18-5
Melting Point -14.0°C
Color Colorless to Yellow
Density 1.2650g/mL
Boiling Point 104.0°C
Flash Point >104°C
Packaging Glass bottle
Molecular Formula C2H2O2
Linear Formula HCOCHO
Quantity 2.5 L
MDL Number MFCD00006957
Beilstein 01, 759
Fieser 01,413
Specific Gravity 1.265
Merck Index 15, 4544
Synonym glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40
Solubility Information Solubility in water: miscible.
InChI Key LEQAOMBKQFMDFZ-UHFFFAOYSA-N
SMILES C(=O)C=O
IUPAC Name oxaldehyde
Molecular Weight (g/mol) 58.04
PubChem CID 7860
Viscosity 8 mPa.s (20°C)
ChEBI CHEBI:34779
Formula Weight 58.04
Percent Purity 39 to 41% (Titrimetry other)
Grade Pure
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.