Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glypican 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577991 Supplier Invitrogen™ Supplier No. PA577991
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: A431, K562, MCF-7, AsPC-1, A431 (24 μg/mL Tunicamycin treatment for 16 hr), A431 (4 mM Benzyl glucosinolate treatment for 32 hr), A431(24 μg/mL Tunicamycin treatment for 16 hr and 4 mM Benzyl glucosinolate treatment for 32 hr) Predicted Reactivity: Mouse (89%), Rat (90%), Rhesus Monkey (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene GPC1
Gene Accession No. P35052
Gene Alias AI462976; glypican; glypican 1; glypican proteoglycan 1; glypican-1; Gpc1; GPI-anchored cell surface heparan sulfate proteoglycan; HSPG M12; HSPGM12; Secreted glypican-1
Gene Symbols GPC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Glypican 1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2817
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.