Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glypican 4 Monoclonal Antibody (965523)
GREENER_CHOICE

Catalog No. MA562007
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA562007 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA562007 Supplier Invitrogen™ Supplier No. MA562007
  • $478.00
    $358.65 / Each of 1

    Save $119.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. The GPC4 gene is adjacent to the 3' end of GPC3 and may also play a role in Simpson-Golabi-Behmel syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 4
Applications ELISA
Classification Monoclonal
Clone 965523
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene GPC4
Gene Accession No. O75487
Gene Alias 9530073D23Rik; AF311610; AI385680; AI661372; dJ900E8.1 (glypican 4); glypican 4; glypican proteoglycan 4; glypican-4; Gpc4; K-glypican; Secreted glypican-4; UNQ474/PRO937
Gene Symbols GPC4
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Glypican 4, Met1-Ser529.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2239
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.