Learn More
Apexbio Technology LLC GM 6001, 10mg. Cas: 142880-36-2 MFCD: MFCD00917040. Broad spectrum MMP inhibitor
SDP
Supplier:  Apexbio Technology LLC A405010
Encompass_Preferred
GM 6001 is a broad-spectrum inhibitor of MMP with Ki values of 0.4nM, 0.5nM, 27nM, 0.1nM and 0.2nM, respectively for MMP-1,2,3,8 and 9 [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4868
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.