Learn More
Invitrogen™ GM-CSF Monoclonal Antibody (DAVKAT), PE, eBioscience™, Invitrogen™
Description
Description: This DAVKAT monoclonal antibody reacts with human Granulocyte-Macrophage Colony-Stimulating Factor (GM-CSF). Human GM-CSF is a 127 amino acid, differentially glycosylated factor produced mainly by activated T cells and macrophages. Endothelial cells and fibroblasts can also produce GM-CSF after exposure to TNF alpha, IL-1, IL-2 and IFN gamma. GM-CSF is found associated with the extracellular matrix and in membrane-bound formats as well. It stimulates proliferation, activation and differentiation of macrophages, granulocytes and their progenitors. Applications Reported: This DAVKAT antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This DAVKAT antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Specifications
Specifications
| Antigen | GM-CSF |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DAVKAT |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CSF2 |
| Gene Accession No. | P04141 |
| Gene Alias | colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.