Learn More
Genscript Corporation Antibody Protein Proteins Proteins 50 UG 1/EA
Supplier:  Genscript Corporation Z0299250
Encompass_Preferred
Granulocyte Macrophage-Colony Stimulating Factor (GM-CSF) was initially characterized as a growth factor that can support the in vitro colony formation of granulocyte-macrophage progenitors. Granulocyte Macrophage-Colony Stimulating Factor (GM-CSF) is produced by a number of different cell types including activated T cells B cells macrophages mast cells endothelial cells and fibroblasts in response to cytokine of immune and inflammatory stimuli. Besides granulocyte-macrophage progenitors Granulocyte Macrophage-Colony Stimulating Factor (GM-CSF) is a growth factor for erythroid megakaryocyte and eosinophil progenitors. On mature hematopoietic monocytes/macrophages and eosinophils. Additionally Granulocyte Macrophage-Colony Stimulating Factor (GM-CSF) can stimulate the proliferation of a number of tumor cell lines including osteogenic sarcoma carcinoma and adenocarcinoma cell lines.
Catalog No. 50-252-9908
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.