Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GM130 Polyclonal Antibody, Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. PIPA1077647 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1077647 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1077647 Supplier Invitrogen™ Supplier No. PA1077A647
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1077A647 detects GM130 from human samples. The PA1077A647 immunogen is a synthetic peptide corresponding to residues G(368) Q V M E S V R Q L Q M E R D K(383) of human GM130.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This encoded protein has been postulated to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GM130
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene Golga2
Gene Accession No. Q08379
Gene Alias 130 kDa cis-Golgi matrix protein; AW555139; cis-Golgi matrix protein GM130; Gm130; GM130 autoantigen; GOLGA2; golgi autoantigen, golgin subfamily a, 2; Golgi matrix protein GM130; golgin A2; Golgin subfamily A member 2; Golgin subfamily A member 2; LOW QUALITY PROTEIN: golgin subfamily A member 2; golgin-95; mKIAA4150; RP11-395P17.5; SY11 protein
Gene Symbols Golga2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues G(368) Q V M E S V R Q L Q M E R D K(383) of human GM130.
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2801
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.