Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GM130 Recombinant Rabbit Monoclonal Antibody (10H5L5), Alexa Fluor™ Plus 647

Catalog No. 703794RP647
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-379-4RP647 50 μL
50-001-02345 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-379-4RP647 Supplier Invitrogen™ Supplier No. 703794RP647
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 658 nm; Emission: 675 nm; Laser: Red Laser.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This encoded protein has been postulated to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GM130
Applications Flow Cytometry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 10H5L5
Concentration 1 mg/mL
Conjugate Alexa Fluor Plus 647
Formulation Proprietary buffer with , and 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.3-7.3
Gene Golga2
Gene Accession No. Q08379
Gene Alias 130 kDa cis-Golgi matrix protein; AW555139; cis-Golgi matrix protein GM130; Gm130; GM130 autoantigen; GOLGA2; golgi autoantigen, golgin subfamily a, 2; Golgi matrix protein GM130; golgin A2; Golgin subfamily A member 2; Golgin subfamily A member 2; LOW QUALITY PROTEIN: golgin subfamily A member 2; golgin-95; mKIAA4150; RP11-395P17.5; SY11 protein
Gene Symbols Golga2
Host Species Rabbit
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2801
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.