Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GMDS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103854
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5103854 100 μL
1 options

Catalog No. PIPA5103854

Supplier: Invitrogen™ PA5103854

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GMDS.

Catalyzes the conversion of GDP-D-mannose to GDP-4-dehydro-6-deoxy-D-mannose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMDS
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GMDS
Gene Accession No. O60547, Q8K0C9
Gene Alias BC031788; C87208; GDP-D-mannose dehydratase; GDP-mannose 4, 6-dehydratase; GDP-mannose 4,6 dehydratase; GDP-mannose 4,6-dehydratase; GMD; GMDS; RP1-118B18.1; SDR3E1; short chain dehydrogenase/reductase family 3E, member 1
Gene Symbols GMDS
Host Species Rabbit
Immunogen A synthesized peptide derived from human GMDS(Accession O60547), corresponding to amino acid residues E277-T327.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218138, 2762, 291095
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.