Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GMEB1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB336641
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336641 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336641 Supplier R&D Systems Supplier No. NBP297215APC

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

GMEB1 Polyclonal specifically detects GMEB1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GMEB1
Applications Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate APC
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias DNA-binding protein p96PIF, glucocorticoid modulatory element binding protein 1, glucocorticoid modulatory element-binding protein 1, GMEB-1, P96PIF, Parvovirus initiation factor p96, PIF p96, PIF96
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GMEB1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10691
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.