Learn More
ACROBiosystems GMP GENIUSNuclease
Supplier:  ACROBiosystems GMPNUES19100KU
Encompass_Preferred
GMP GENIUS™Nuclease is a tag free recombinant form of Serratia marcescens extracellular endonuclease produced in Escherichia coli cells using a proprietary process at ACRObiosystems. GMP GENIUSNuclease DMF Filed is a homodimer with monomer molecular masses about 30 kDa with an isoelectric point (pI) at pH 6.85. The enzyme is a non-specific nuclease with high specific activity which degrades both single- and double-stranded nucleic acids in any form (single stranded double stranded linear circular and supercoiled). It hydrolyzes internal phosphodiester bonds present between the nucleotides to 5’- phosphorylated oligonucleotides of 3-5 bases in length.
Catalog No. 50-253-4969
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.