Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GMP Human AB Serum
SDP

Catalog No. RD10008956 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10008956 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10008956 Supplier R&D Systems™ Supplier No. HABS001GMP100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GMP grade media supplement for cell therapy workflows.

/////Used to formulate 2 liters of GMP Human T Cell Media at 5% supplementation/////cGMP, Xeno-free, and testing is compliant with requirements outlined in 21 CFR 610.40/////Extensive viral testing done at the donor and pool levels/////Small donor pool derived material///// Heat inactivation protocol available///// Have questions? See our FAQs

Specifications

Content And Storage Store the unopened product frozen at ≤ -10°C. See Use by Date on the Certificate of Analysis.
Quantity 100 mL
Source Human
Product Line R&D Systems
Regulatory Status GMP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.