Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GMP Human T Cell Media
SDP

Catalog No. CCM038GMP1B Shop All Novus Products
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CCM038GMP1B 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CCM038GMP1B Supplier R&D Systems™ Supplier No. CCM038GMP1B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For robust T cell expansion with GMP-grade media in xeno-free conditions

/////Phenol Red-Free formulation in 1 L bottles (CCM038-GMP-1L) or 1 L bags (CCM038-GMP-1B)/////Robust T cell expansion under xeno-free conditions/////Optimized for use with Bio-Techne cytokines and G-Rex™ bioreactors/////Compatible with a variety of T cell activation methods/////Supports T cell expansion from PBMCs

Specifications

Product Line R&D Systems
Quantity 1000 mL
Species Human
Content And Storage Store the unopened product at 2-8 °C and protected from light. Do not use past the use by date on the Certificate of Analysis.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.