Learn More
ACROBiosystems GMP Human TNF-alpha Protein
Supplier:  ACROBiosystems GMPTNAH231MG
Encompass_Preferred
GMP Human TNF-alpha Protein (GMP-TNAH23) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P01375).
Catalog No. 50-253-4963
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.