Learn More
Compass Biomedical™ GMP PLUS™ Human Platelet Lysate

Description
- Potent: Human derived product rich in growth factors, cytokine, and other proteins
- Products are growth factor rich supplements made from large pools of transfusion grade human platelets sourced from FDA-registered/AABB accredited blood centers.
Specifications
Specifications
| For Use With (Application) | Cell Culture, Stem Cell Culture |
| Quantity | 500 mL |
| Purity | GMP Grade |
| Host Species | Human |
| Product Type | Human Platelet Lysate |
| Content And Storage | Store at -80°C upon receipt. Thaw at room temperature or in a water bath. After thawing, aliquot and store at -20°C or colder. Long term storage at -80°C is recommended. Multiple freeze/thaw cycles are not recommended. |
| Format | Bottle |
| Research Category | GMP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.