Learn More
Cytiva GN Metricel MCE Membrane Disc Filters - 25 mm, grid (100/pkg)
Supplier:  Cytiva 64191
Encompass
GN Metricel MCE Membrane Disc Filters - 25 mm, grid (100/pkg) Material:Hydrophilic mixed cellulose esters; Maximum Operating Temperature (Metric)74 °C; Typical Filter Weight (Metric) 4 mg/cm²; Water Flow Rate65 mL/min/cm²
Catalog No. 50-206-3270
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.