Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNAI1/GNAI2/GNAI3 Recombinant Rabbit Monoclonal Antibody (HL2092)
GREENER_CHOICE

Catalog No. FEMA550378
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550378 Supplier Invitrogen™ Supplier No. MA550378
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

GNAI1/GNAI2/GNAI3 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

GNAI1/GNAI2/GNAI3 are genes that encode for the G protein alpha inhibiting activity subunit. These proteins play a crucial role in signal transduction pathways, particularly in the inhibition of adenylyl cyclase. They are involved in various physiological processes, including neurotransmitter release, hormone secretion, and regulation of smooth muscle contraction. Mutations in these genes have been associated with various diseases, including hypoparathyroidism, pituitary adenoma, and cardiovascular disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNAI1/GNAI2/GNAI3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2092
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GNAI3
Gene Accession No. B2RSH2, P04897, P04899, P08752, P08753, P08754, P10824, P63096, Q9DC51
Gene Alias HG1B/HG1C/HG1A
Gene Symbols GNAI1, GNAI2, GNAI3
Host Species Rabbit
Immunogen Recombinant fragment of human GNAI3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100006888, 14677, 14678, 14679, 25643, 25686, 2770, 2771, 2773, 393946, 81664
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.