Learn More
Medchemexpress LLC HY-12947 10mg Medchemexpress, GNE-3511 CAS:1496581-76-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1294710MG
Encompass_Preferred
Medchemexpress, HY-12947 10mg GNE-3511 CAS:1496581-76-0 GNE-3511 is a dual leucine zipper kinase (DLK) inhibitor with a Ki of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1208
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.