Learn More
Cayman Chemical GNE-3511, 1496581-76-0, 1 mg
SDP
Supplier:  Cayman Chemical 191741
Encompass_Preferred
A selective and highly potent inhibitor of DLK; also inhibits phosphorylated JNK (IC50 = 30 nM); selective over other MAP kinases, JNKs, and MLKs; protects primary neurons in an in vitro axon degeneration assay (IC50 = 107 nM); decreases p-c-Jun expression in a mouse MPTP model of Parkinson’s disease,
Catalog No. NC1386280
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.