Learn More
Selleck Chemical LLC GNF351
SDP
Supplier:  Selleck Chemical LLC S69265MG
Encompass_Preferred
GNF351 is a potent antagonist of aryl hydrocarbon receptor (AHR) GNF351 interacts directly with the AHR ligand binding pocket and competes with a well-characterized photoaffinity AHR ligand for binding to the AHR with IC50 of 62 nM
Catalog No. 50-217-3238
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.