Learn More
Cayman Chemical GnIrelIxacetate 10mg
SDP
Supplier:  Cayman Chemical 2409810 MG
Encompass_Preferred
A GNRHR antagonist (IC50 = 3.6 nM; pA2 = 9.3); induces a concentration-dependent increase in histamine release from rat peritoneal mast cells in vitro (EC50 = 11 μg/ml); decreases plasma testosterone in intact male rats for the first 7 days post-administration (2 mg/kg, s.c.); decreases the surface area of endometriotic lesions and serum progesterone levels in female baboons (125 μg per day for 30 days)
Catalog No. 50-293-4119