Learn More
Bachem (Des-Gly10,D-Ala6,Pro-NHEt9)-LHRH (salmon)
SDP
Supplier:  Bachem 4013835.0001
Encompass_Preferred
(Des-Gly10,D-Ala6,Pro-NHEt9)-LHRH (salmon), Pyr-His-Trp-Ser-Tyr-D-Ala-Trp-Leu-Pro-NHEt, 88848-87-7, C61H76N14O12, Mr 1197.36, 1mg. GnRH analog used in aquaculture.
Catalog No. 50-194-6988
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.