Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GnRH1 Monoclonal Antibody (LHRH13-387.5), Invitrogen™
Mouse Monoclonal Antibody
$450.00 - $1108.00
Specifications
Antigen | GnRH1 |
---|---|
Clone | LHRH13-387.5 |
Concentration | 2.4 mg/mL |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA546178
|
Thermo Scientific
MA546178 |
100μg |
Each for $450.00
|
|
|||||
01-671-844
|
Thermo Scientific
MA110816 |
500 μg |
Each for $572.50
|
|
|||||
PIMA546177
|
Thermo Scientific
MA546177 |
1 mg |
Each for $1,108.00
|
|
|||||
Description
MA1-10816 detects Gonadotropin Releasing Hormone from human samples. MA1-10816 has been successfully used in ELISA and immunohistochemistry applications. The MA1-10816 immunogen is purified human gonadotropin releasing hormone.
The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).Specifications
GnRH1 | |
2.4 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1 | |
GNRH1 | |
IgG1 | |
Protein A | |
Antibody |
LHRH13-387.5 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P01148 | |
2796 | |
Purified human gonadotropin releasing hormone. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
GNRH1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title