Learn More
Description
PA1-121 detects gonadotropin releasing hormone (GnRH) in mouse brain and in amphibian (Hyla cinerea) samples. PA1-121 has been successfully used in immunohistochemistry procedures. Immunohistochemical staining of GnRH in mouse embryo brain tissue using PA1-121 results in staining of neurons in the organum vasculosum of the lamina terminalis (OVLT). In immunohistochemical procedures, 4% paraformaldehyde is recommended as a fixative. The PA1-121 immunogen is a synthetic peptide corresponding to residues pyroE(22) H W S Y G L R P G-NH2(32) of mouse GnRH. This sequence is completely conserved in rat and tree shrew and is 78% conserved in zebrafish. The PA1-121 immunizing peptide (Cat. # PEP-168) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | GnRH1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA and 0.05% sodium azide |
| Gene | GNRH1 |
| Gene Accession No. | P13562 |
| Gene Alias | Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1 |
| Gene Symbols | GNRH1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
