Learn More
Vernier™ Go Direct™ Charge Station
SDPSCI_ED
Catalog No. S39706 Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Go Direct Sensors, Go Wireless Sensors
Catalog No. For Use With (Equipment)
S39706 Go Direct Sensors, Go Wireless Sensors
1 options
Catalog No. S39706 Supplier Vernier™ Supplier No. GDXCRG
Add to Cart
Add to Cart
Vernier™ Go Direct™ Charge Station for charging Go Direct™ Sensors and Go Wireless Sensors.
  • Each charge station has sixteen charging ports - eight USB and eight wand-style sensor ports.
  • LED lights on the Go Direct™ sensors will show charging status.
  • Also compatible with Go Wireless devices.
  • USB cables not included.
Specifications
Includes 16 charging ports
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Go Direct Sensors, Go Wireless Sensors