Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go Direct™ Energy Sensor
SDPSCI_ED

Catalog No. S39721 Shop All Vernier Software Products
Change view
Click to view available options
Grade:
K-8, High School, College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S39721 K-8, High School, College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39721 Supplier Vernier™ Supplier No. GDXNRG
Add to Cart
Add to Cart

Vernier™ Go Direct™ Energy Sensor quantifies the voltage, current, power, and energy output of small wind turbines and solar panels, such as those used in KidWind Experiment Kits.

  • Investigate the electrical energy generated by a wind turbine or solar panel.
  • Explore the effect of load on wind turbine or solar panel output.
  • Test blade design variables and evaluate data to determine optimal blade design.
  • Source input potential range: ±30 V
  • Source input current range: ±1 A

Specifications

Grade K-8, High School, College
Interface Bluetooth or USB
Warranty 5 Year Limited
For Use With (Application) Graphical Analysis 4 or LabQuest App
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.