Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go Direct™ Gas Pressure Sensor
SDPSCI_ED

Catalog No. s39713 Shop All Vernier Software Products
Change view
Click to view available options
Range:
0 to 400 kPa
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S39713 0 to 400 kPa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39713 Supplier Vernier™ Supplier No. GDXGP
Only null left
Add to Cart
Add to Cart

Vernier™ Go Direct™ Gas Pressure Sensor measures the absolute pressure of a gas.

  • Maximum sampling rate: 50 samples/s
  • Internal volume: 0.8 mL
  • Maximum pressure that the sensor can tolerate without permanent damage: 450 kPa
  • Investigate Boyle's law and Charles' law
  • Investigate grip strength and muscle fatigue (using Gas Pressure Bulb, not included)
  • Measure the production of oxygen gas produced when hydrogen peroxide is destroyed by the enzyme catalase
  • Monitor plant transpiration

Specifications

Grade K-8, High School, College
Interface Bluetooth or USB
Warranty 5 Year Limited
For Use With (Application) Graphical Analysis 4 or LabQuest App
Accuracy ±3 kPa
Range 0 to 400 kPa
Resolution 0.03 kPa
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.