Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go Direct™ Glass-Body pH Sensor
SDPSCI_ED

Catalog No. S39714 Shop All Vernier Software Products
Change view
Click to view available options
Temperature Range (Metric):
0°C to 80°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
S39714 0°C to 80°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39714 Supplier Vernier™ Supplier No. GDXGPH
Add to Cart
Add to Cart

Vernier™ Go Direct™ Glass-Body pH Sensor for measuring the pH in aqueous, heterogeneous, and organic solutions.

  • This high-quality glass body pH sensor can be used in non-aqueous solutions and solutions that contain organic solvents, strong acids, or strong bases. The electrode features a sealed, gel-filled Ag-AgCl combination reference electrode and uses a BNC connector.
  • Accuracy: ±0.2 pH units (factory calibration), ±0.05 pH units (user calibration)
  • Response time: 2 s (to 90% of full reading in aqueous buffer), 5 s (to 90% of full reading in 50% acetonitrile)
  • 12 mm OD

Specifications

Includes Glass shaft, sealed combination electrode with Ag/AgCl reference
Grade High School and College
Interface Bluetooth or USB
Material Glass
Warranty 5 Year Limited
For Use With (Application) Graphical Analysis 4 or LabQuest App
Range 0 to 14 pH
Temperature Range (Metric) 0°C to 80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.