Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go Direct™ Motion Detector
SDPSCI_ED

Catalog No. S39717 Shop All Vernier Software Products
Change view
Click to view available options
Range:
15 cm to 3.5 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S39717 15 cm to 3.5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39717 Supplier Vernier™ Supplier No. GDXMD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vernier™ Go Direct™ Motion Detector uses ultrasound to measure the position, velocity, and acceleration of moving objects.

  • Accurately tracks objects as close as 15 cm and as far away as 3.5 m. The compact design and wireless capability of this motion detector eliminate the concern of a dangling cable getting in the way.
  • Study position, velocity, and acceleration of carts when mounted on a track.
  • Match graphs created in Graphical Analysis 4 software.
  • Analyze the effects of air resistance on falling coffee filters.
  • Investigate simple harmonic motion by monitoring a mass on a spring.

Specifications

Grade K-8, High School, College
Interface Bluetooth or USB
Warranty 5 Year Limited
For Use With (Application) Graphical Analysis 4 or LabQuest App
Range 15 cm to 3.5 m
Rate 30 samples/s
Resolution 1 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.