Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go Direct™ ORP Sensor
SDPSCI_ED

Catalog No. S39724 Shop All Vernier Software Products
Change view
Click to view available options
Temperature Range (Metric):
0°C to 60°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
S39724 0°C to 60°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39724 Supplier Vernier™ Supplier No. GDXORP
Add to Cart
Add to Cart

Vernier™ Go Direct™ ORP Sensor measures the ability of a solution to act as an oxidizing or reducing agent.

  • Use the Go Direct ORP Sensor to measure the ability of a solution to act as an oxidizing agent and to quantify ion activity. This sensor measures the capacity of a solution to either release or accept electrons from chemical reactions.
  • This sensor is comprised of an ORP electrode connected to a Go Direct Electrode Amplifiervia a BNC connector. This design enables you to use the Go Direct Electrode Amplifier with other Vernier electrodes, such as the pH Electrode, Flat pH Electrode, ORP Electrode, ISE electrode, or a third-party electrode.
  • Determine the equivalence point of an oxidation-reduction titration.
  • Measure the oxidizing ability of chlorine in swimming pools.
  • Investigate the ORP of drinking water.
  • Storage solution: pH-4/KCl solution (10 g KCl in 100 mL buffer pH-4 solution)
  • ORP element: 99% pure platinum band sealed on a glass stem
  • 12 mm OD
  • Impedance: ∼20 kΩ at 25°C
  • Input range: ±1000 mV

Specifications

Type Sealed, Gel-filled, Epoxy Body, Ag/AgCl Reference
Includes Micro USB Cable
Grade High School and College
Interface Bluetooth or USB
Warranty 5 Year Limited
For Use With (Application) Graphical Analysis 4 or LabQuest App
Accuracy ±20 mV
Temperature Range (Metric) 0°C to 60°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.