Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go Direct™ pH Sensor
SDPSCI_ED

Catalog No. s39725 Shop All Vernier Software Products
Change view
Click to view available options
Range:
0 to 14 pH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S39725 0 to 14 pH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39725 Supplier Vernier™ Supplier No. GDXPH
Only null left
Add to Cart
Add to Cart

Vernier™ Go Direct™ pH Sensor is a general-purpose pH sensor used to monitor pH of aqueous solutions.

  • Storage solution: pH-4/KCl solution (10 g KCl in 100 mL buffer pH-4 solution)
  • The Go Direct pH Sensor is an important and versatile sensor for lab and field activities alike. It gives students the freedom to explore pH without the inconvenience of wires—no more spilled solutions or dangling wires. The Go Direct pH transmits live readings and captures data in real-time.
  • Conduct acid-base titrations.
  • Monitor pH change during chemical reactions.
  • Test the pH and alkalinity of bodies of water.
  • Investigate household acids and bases.
  • This sensor is comprised of a pH electrode connected to a Go Direct Electrode Amplifier via a BNC connector. This design enables you to use the Go Direct Electrode Amplifier with other Vernier electrodes, such as the Flat pH Electrode, ORP Electrode, ISE electrode, or a third-party electrode.

Specifications

Type Sealed, Gel-filled, Epoxy Body, Ag/AgCl Reference
Description 12 mm OD
Grade Middle School, High School, College
Interface Bluetooth or USB
Warranty 5 Year Limited
For Use With (Application) Graphical Analysis 4 or LabQuest App
Accuracy ±0.2 pH
Range 0 to 14 pH
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.