Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go Direct™ Radiation Monitor
SDPSCI_ED

Catalog No. S39727 Shop All Vernier Software Products
Change view
Click to view available options
Grade:
High School and College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S39727 High School and College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39727 Supplier Vernier™ Supplier No. GDXRAD
Add to Cart
Add to Cart

Vernier™ Go Direct™ Radiation Monitor detects alpha, beta, gamma, and X-ray radiation.

  • Sensor: LND 712 (or equivalent) halogen-quenched GM tube with a mica end window, 1.5 to 2.0 mg/cm2 thick
  • Sensitive to alpha, beta, gamma, and X-ray radiation
  • Powered by rechargeable 300 mAh battery, recharged via USB cable
  • Red LED blinks on radiation detection
  • Optional audio signal on radiation detection
  • Use the Go Direct Radiation Monitor to explore radiation statistics, measure the rate of nuclear decay, and monitor radon progeny. This easy-to-use sensor consists of a Geiger-Mueller tube mounted in a small, rugged, plastic case. A thin window protected by a metal screen allows alpha radiation to be detected, along with beta and gamma. Each new detection is indicated by a flashing red LED indicator and can also be accompanied by an optional audio signal.

Specifications

Grade High School and College
Interface Bluetooth or USB
Warranty 5 Year Limited
For Use With (Application) Graphical Analysis 4 or LabQuest App
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.