Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go Direct™ Tris-Compatible Flat pH Sensor
SDPSCI_ED

Catalog No. S39712 Shop All Vernier Software Products
Change view
Click to view available options
Temperature Range (Metric):
0°C to 100°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
S39712 0°C to 100°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39712 Supplier Vernier™ Supplier No. GDXFPH
Add to Cart
Add to Cart

Vernier™ Go Direct™ Tris-Compatible Flat pH Sensor for measuring the pH of a solution or semisolid, such as food or soil.

  • Storage solution: pH 4/KCl solution (10 g KCl in 100 mL buffer pH-4 solution)
  • 12 mm OD
  • Impedance: ∼20 kΩ at 25°C
  • Glass membrane is flat instead of a bulb, making it more durable, easier to clean, and allows for flat surface measurements or smaller sample sizes.
  • It features a sealed, gel-filled, double-junction electrode, making it compatible with Tris buffers and solutions containing proteins or sulfides.
  • The flat glass shape also makes it useful for measuring the pH of semisolids such as food or soil slurries.

Specifications

Description Flat glass membrane style
Includes Double-junction, sealed, gel-filled, Ag/AgCl reference, polycarbonate body electrode
Grade High School and College
Interface Bluetooth or USB
Warranty 5 Year Limited
For Use With (Application) Graphical Analysis 4 or LabQuest App
Range 0 to 14 pH
Response Time 98% of full response in 30 seconds at 25°C
Temperature Range (Metric) 0°C to 100°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.