Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Go Direct™ Weather Vane
SDPSCI_ED

Catalog No. S1724330 Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Go Direct™ weather sensor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S1724330 Go Direct™ weather sensor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S1724330 Supplier Vernier Supplier No. WTHRVANE
Add to Cart
Add to Cart

The Go Direct™ weather vane accessory is required for Go Direct™ weather (GDX-WTHR) to report the direction of the wind.

Includes

  • Go Direct™ Weather Vane*

*Go Direct™ Weather (GDX-WTHR) & Tripod - not included

Specifications

Grade High School and College
For Use With (Application) Used to report the direction of the wind
For Use With (Equipment) Go Direct™ weather sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.