Learn More
Vernier™ Go!Link Teacher Pack
SDPSCI_ED
Catalog No. S16157ND Shop All Vernier Software Products
Change view
Click to view available options
Warranty:
5 Year Limited
Catalog No. Warranty
S16157ND 5 Year Limited
1 options
Catalog No. S16157ND Supplier Vernier™ Supplier No. GLTP
Add to Cart
Add to Cart
Includes
Go!Link Teacher Pack
Use to connect 8 sensors to 8 computers or connect multiple sensors to one computer or Chromebook.
  • USB specification: 1.1
  • Maximum sample rate: 200 samples per second
  • a Qquick and affordable way to get started with data-collection technology.
  • It's a single-channel interface that connects most Vernier sensors to your computer or Chromebook USB port.
Warranty and Services
5-year limited warranty
Specifications
Includes 8 Go!Links
Warranty 5 Year Limited
For Use With (Application) Educational use only
Resolution 12 bit