Learn More
Vernier™ Go!Motion to Calculator Cable
SDPSCI_ED
Catalog No. S16158ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
CBR 2 and Go!Motion
Catalog No. For Use With (Equipment)
S16158ND CBR 2 and Go!Motion
1 options
Catalog No. S16158ND Supplier Vernier™ Supplier No. GMCALC
Includes
Go!Motion to Calculator Cable
This 84? cable is used to connect a Go!Motion or CBR 2 to the I/O port of a TI graphing calculator. The cable has the small I/O plug on both ends. The cable can also be used to connect a graphing calculator to the I/O port on a LabPro, CBL 2, or another calculator. The cable is included with a CBR 2.
  • The cable has the small I/O plug on both ends.
  • The cable can also be used to connect a graphing calculator to the I/O port on a LabPro, CBL 2, or another calculator.
  • The cable is included with a CBR 2.
Warranty and Services
5-year limited warranty
Specifications
Warranty 1-year limited warranty from Texas Instruments
For Use With (Application) Educational use only
For Use With (Equipment) CBR 2 and Go!Motion
Length (Metric) 2.13 m (Cable)