Learn More
Vernier™ Go!Motion to Computer Cable
SDPSCI_ED
Catalog No. S16159ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
CBR 2 and Go!Motion
Catalog No. For Use With (Equipment)
S16159ND CBR 2 and Go!Motion
1 options
Catalog No. S16159ND Supplier Vernier™ Supplier No. GMCUSB
Add to Cart
Add to Cart
Includes
Go!Motion to Computer Cable
This cable allows a Go!Motion or CBR 2 to connect to a computer. It is included with Go!Motion.
  • This cable allows a Go!Motion or CBR 2 to connect to a computer.
  • It is included with Go!Motion.
  • This cable will not work for spectrometers or LabPro. A LabPro USB Cable is needed instead.
Warranty and Services
5-year limited warranty
Specifications
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) CBR 2 and Go!Motion