Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go!Motion
SDPSCI_ED

Catalog No. S16162ND Shop All Vernier Software Products
Change view
Click to view available options
Range:
0.15 to 6 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S16162ND 0.15 to 6 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16162ND Supplier Vernier™ Supplier No. GOMOT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Go!Motion, Vernier Go!Motion, Standard USB cable, User manual, Go!Motion Teacher Pack, 8 Vernier Go!Motion, 8 Standard USB cables, User manual

Requires

No interface required and free data-collection software download

Go!Motion is our motion detector that connects directly to a computer or Chromebook USB port—eliminating the need for an additional data-collection interface.

Range: 0.15 to 6 m Resolution: 1 mm Temperature Compensation: Automatic Accuracy: 2mm Ultrasound Frequency: 50 kHz Power Consumption: 51 mA Power Source: USB/DIG (Computers, LabQuest) or 4 AA batteries (Calculators)

Warranty and Services

5-year limited warranty

Specifications

Warranty 5 Year Limited
For Use With (Application) Educational use only
Accuracy 2 mm
Range 0.15 to 6 m
Resolution 1 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.