Learn More
Vernier™ Go Wireless®Bluetooth®Adapter
SDPSCI_ED
Catalog No. S16171ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
LabQuest 2
Catalog No. For Use With (Equipment)
S16171ND LabQuest 2
1 options
Catalog No. S16171ND Supplier Vernier™ Supplier No. GWRADIO
Includes
Go Wireless®Bluetooth® Adapter
The Go Wireless® Bluetooth Adapter enables older models of LabQuest 2 to wirelessly communicate with Go Direct sensors wirelessly.
  • For use with LabQuest 2 units (Model: LABQUEST2) running LabQuest app software version 2.5 or newer.
Warranty and Services
5-year limited warranty
Specifications
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) LabQuest 2