Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go Wireless™Exercise Heart Rate
SDPSCI_ED

Catalog No. S16166ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
Middle School, High School, College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16166ND Middle School, High School, College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16166ND Supplier Vernier™ Supplier No. GWEHR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Exercise Heart Rate Strap, Polar Transmitter Module

Requires

Requires an interface and software, sold separately

The Vernier Go Wireless™Exercise Heart Rate is an excellent hands-free option for continuously monitoring heart rate before, during, and after exercise or while a person is stationary. Data are wirelessly transmitted to iPad™, LabQuest 2, or Android™devices.

Battery Type: CR 2025 Battery Lifetime: 200 hrs Operating Temperature: -10 to 50 °C Wireless range: 10 m or more unobstructed Radios: Bluetooth and 5 kHz RF transmission

Warranty and Services

5-year limited warranty

Specifications

Grade Middle School, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Operating Temperature (Metric) -10°C to +50°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.