Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Alligator Whole Serum Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NB10055303 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10055303 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10055303 Supplier Novus Biologicals Supplier No. NB10055303
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Whole Serum Polyclonal specifically detects Whole Serum in Alligator samples. It is validated for Immunoelectrophoresis.

Specifications

Antigen Whole Serum
Applications Immunoelectrophoresis
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunoelectrophoresis
Host Species Goat
Immunogen Alligator Whole Serum
Purification Method Affinity Purified
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Alligator.
Target Species Alligator
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.