Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Bat IgG (H+L) Secondary Antibody, Biotin, R&D Systems™
SDP

Catalog No. NBP222358 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222358 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222358 Supplier R&D Systems Supplier No. NBP222358
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Bat samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Dilution Western Blot 1;20000-1:400000, ELISA 1:20000-1:400000, Immunohistochemistry 1:500-1:5000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
Host Species Goat
Immunogen Bat IgG-heavy and light chain
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this antibody reacts specifically with Bat IgG and with light chains common to other Bat immunoglobulins. Antibody has been shown to react with bat genus species Pteropus vampirus, Desmodus rotundus, Eptesicus fuscus, Tadrida pumila, T. condylura, Hypsignathus monstrosus, Rosettus aegyptiacus, Epomorphus crypturus, Molossus species, and Phylostomus species. No antibody was detected against non-immunoglobulin serum proteins. This may cross react with IgG from other species.
Target Species Bat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.