Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Goat anti-Bovine IgG (H+L) Secondary Antibody, Agarose

Catalog No. PA130750
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA130750 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA130750 Supplier Invitrogen™ Supplier No. PA130750

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Goat Polyclonal Secondary Antibody

PA1-30750 detects Bovine IgG from bovine samples.

Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bovine IgG (H+L)
Applications Affinity Purification
Classification Polyclonal
Concentration 9.2 mg/mL
Conjugate Agarose
Formulation PBS with 0.01% sodium azide; pH 7.2
Host Species Goat
Immunogen Bovine IgG whole molecule.
Purification Method IgG fraction
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Slurry
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.