Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Canine IgE Heavy Chain Secondary Antibody, DyLight 594, R&D Systems™
SDP

Catalog No. NB540408 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB540408 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB540408 Supplier R&D Systems Supplier No. NB7343DL594
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgE Heavy Chain Polyclonal specifically detects IgE Heavy Chain in Canine samples. It is validated for Dot Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgE Heavy Chain
Applications Dot Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon
Host Species Goat
Immunogen Dog IgE
Purification Method Immunogen affinity purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Canine
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.