Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Canine IgG Fc Secondary Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NBP1733501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1733501 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP1733501 Supplier Novus Biologicals Supplier No. NBP1733501MG
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Canine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.

Specifications

Antigen IgGFc
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate FITC
Dilution Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Formulation Lyophilized from 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2, with 10Mg/ml Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Host Species Goat
Immunogen Dog IgG Fc fragment
Purification Method Affinity Purified
Quantity 1 mg
Primary or Secondary Secondary
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Goat Serum, Dog IgG, Dog IgG F(c) and Dog Serum. No reaction was observed against Dog IgG F(ab).
Reconstitution Reconstitute with 1.0mL of deionized water.
Target Species Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.