Learn More
Goat anti-Canine IgG (H+L) Secondary Antibody, Allophycocyanin, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunomicroscopy |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Host Species | Goat |
| Immunogen | This Goat anti-Canine IgG (H+L) Secondary Antibody was developed against purified canine IgG (H&L). |
| Purification Method | Affinity purified |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.